Nicotinic acid, 99%
Nicotinic acid, 99%
Nicotinic acid, 99%
Thermo Scientific Chemicals

Nicotinic acid, 99%

CAS: 59-67-6 | C6H5NO2 | 123.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A12683.0B
also known as A12683-0B
Price (USD)
104.00
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
104.00
Each
Chemical Identifiers
CAS59-67-6
IUPAC Namepyridine-3-carboxylic acid
Molecular FormulaC6H5NO2
InChI KeyPVNIIMVLHYAWGP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥98.5% (UK-sourced material)
CommentMaterial sourced in UK and US
Melting Point (clear melt)234.0-241.0°C
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5% (UK-sourced material)
View more
Vitamin of the B complex with hypolipidemic properties occurring in various animal and plant tissues. Required by the body for the formation of coenzymes NAD and NADP.Nicotinic acid is used as an antidyslipidemic agent. It is also used to improve all lipoproteins well above the vitamin requirement and lowers the total cholesterol, bad LDL-cholesterol, and triglyceride levels. Further, it is used as a food additive in food industries, cosmetics and pharmaceuticals. In addition to this, it is used in medication, nutrient, enriched flours and in electroplating baths.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water. Slightly soluble in ethanol and ethyl ether. Insoluble in most lipid solvents.

Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Cheng, X.; Hookway, E. S.; Kashima, T.; Oppermann, U.; Galione, A.; Athanasou, N. A. The Role of Calcium and Nicotinic Acid Adenine Dinucleotide Phosphate (NAADP) in Human Osteoclast Formation and Resorption. Calcif. Tissue Int. 2015, 96 (1), 73-79.
  2. Trabbic, C. J.; Zhang, F.; Walseth, T. F.; Slama, J. T. Nicotinic Acid Adenine Dinucleotide Phosphate Analogues Substituted on the Nicotinic Acid and Adenine Ribosides. Effects on ReceptorMediated Ca2+ Release. J. Med. Chem. 2015, 58 (8), 3593-3610.