(-)-Borneol, 97+%
(-)-Borneol, 97+%
(-)-Borneol, 97+%
Thermo Scientific Chemicals

(-)-Borneol, 97+%

CAS: 464-45-9 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A12684.0B
also known as A12684-0B
Price (USD)
416.65
Online Exclusive
463.00
Save 46.35 (10%)
Each
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)
416.65
Online Exclusive
463.00
Save 46.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS464-45-9
IUPAC Name(1S,2R,4R)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-ol
Molecular FormulaC10H18O
InChI KeyDTGKSKDOIYIVQL-NQMVMOMDSA-N
SMILESCC1(C)[C@@H]2CC[C@]1(C)[C@H](O)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.0%
Identification (FTIR)Conforms
Optical Rotation-30.0 ± 10.0° (C=5 in ethanol)
Appearance (Color)White
FormCrystals or powder or crystalline powder or lumps
(-)-Borneol is used to prepare its esters by reacting with acids. Its derivatives are used as chiral ligands in asymmetric synthesis. It is also used in flavors and perfumes. Further, it is used in traditional Chinese medicine as moxa. In addition to this, it is used as a component of many essential oils and also used as a natural insect repellent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-Borneol is used to prepare its esters by reacting with acids. Its derivatives are used as chiral ligands in asymmetric synthesis. It is also used in flavors and perfumes. Further, it is used in traditional Chinese medicine as moxa. In addition to this, it is used as a component of many essential oils and also used as a natural insect repellent.

Solubility
Soluble in chloroform, ethanol, acetone, ether, benzene, toluene, decalin and tetralin. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Christoforides, E.; Mentzafos, D.; Bethanis, K. Structural studies of the inclusion complexes of the (+)- and (-)-borneol enantiomers in alpha- and beta-cyclodextrin. J. Incl. Phenom. Macro. Chem. 2015, 81 (1), 193-203.
  2. Marumoto, S.; Okuno, Y.; Miyamoto, Y.; Miyazawa, M. Biotransformation of (+)-(1R,2S,4R)-borneol and (-)-(1S,2R,4S)-borneol by Spodoptera litura (common cutworm) larvae. J. Mol. Catal. B: Enzym. 2015, 115, 160-167.