5-Chloro-3-methyl-1-phenyl-1H-pyrazole, 98%
5-Chloro-3-methyl-1-phenyl-1H-pyrazole, 98%
5-Chloro-3-methyl-1-phenyl-1H-pyrazole, 98%
Thermo Scientific Chemicals

5-Chloro-3-methyl-1-phenyl-1H-pyrazole, 98%

CAS: 1131-17-5 | C10H9ClN2 | 192.646 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A12685.06
also known as A12685-06
Price (USD)
234.65
Online Exclusive
261.00
Save 26.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
234.65
Online Exclusive
261.00
Save 26.35 (10%)
Each
Chemical Identifiers
CAS1131-17-5
IUPAC Name5-chloro-3-methyl-1-phenyl-1H-pyrazole
Molecular FormulaC10H9ClN2
InChI KeyZZOWFLAMMWOSCG-UHFFFAOYSA-N
SMILESCC1=NN(C(Cl)=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5765-1.5805 @ 20?C
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to yellow
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only