2-Bromo-3-hydroxypyridine, 99%
2-Bromo-3-hydroxypyridine, 99%
2-Bromo-3-hydroxypyridine, 99%
Thermo Scientific Chemicals

2-Bromo-3-hydroxypyridine, 99%

CAS: 6602-32-0 | C5H4BrNO | 174.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12687.14
also known as A12687-14
Price (USD)
191.65
Online Exclusive
213.00
Save 21.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
191.65
Online Exclusive
213.00
Save 21.35 (10%)
Each
Chemical Identifiers
CAS6602-32-0
IUPAC Name2-bromopyridin-3-ol
Molecular FormulaC5H4BrNO
InChI KeyYKHQFTANTNMYPP-UHFFFAOYSA-N
SMILESOC1=CC=CN=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)180.0-187.0?C
Appearance (Color)White to pale cream
Assay (GC)≥98.5%
2-Bromo-3-hydroxypyridine is used to produce 2-bromo-4,6-diiodo-3-pyridinol. It is also used in azo dyes and in the synthesis of pterocellin A by reacting with kojic acid and in synthesis of orelline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-3-hydroxypyridine is used to produce 2-bromo-4,6-diiodo-3-pyridinol. It is also used in azo dyes and in the synthesis of pterocellin A by reacting with kojic acid and in synthesis of orelline.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Parhi, A. K.; Xiang, A.; Bauman, J. D.; Patela, D.; Vijayan, R. S. K.; Das, K.; Arnold, E.; LaVoie, E. J. Phenyl substituted 3-hydroxypyridin-2(1H)-ones: Inhibitors of influenza A endonuclease. Bioorg. Med. Chem. 2013, 21 (21), 6435-6446.
  2. Ganguly, N. C.; Mondal, P.; Roy, S.; Mitra, P. Ligand-free copper-catalyzed efficient one-pot access of benzo[b]pyrido[3,2-f][1,4]oxazepinones through O-heteroarylation-Smiles rearrangement-cyclization cascade. RSC Adv. 2014, 4 (98), 55640-55648.
  3. Kato, Y.; Kawasaki, M.; Nigo, T.; Nakamur, S.; Fusano, A.; Teranishi, Y.; Ito, M. N.; Sumiyoshi, T. Identification of 2,3-disubstituted pyridines as potent, orally active PDE4 inhibitors. Bioorg. Med. Chem. 2013, 21 (18), 5851-5854.