2-Acetylpyridine, 98%
2-Acetylpyridine, 98%
2-Acetylpyridine, 98%
Thermo Scientific Chemicals

2-Acetylpyridine, 98%

CAS: 1122-62-9 | C7H7NO | 121.139 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A12688.14
also known as A12688-14
Price (USD)
44.65
Online Exclusive
49.50
Save 4.85 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
44.65
Online Exclusive
49.50
Save 4.85 (10%)
Each
Chemical Identifiers
CAS1122-62-9
IUPAC Name1-(pyridin-2-yl)ethan-1-one
Molecular FormulaC7H7NO
InChI KeyAJKVQEKCUACUMD-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5220 - 1.5240 @20?C
Appearance (Color)Clear, colourless to pale yellow
Assay (GC)> 97.5%
Formliquid
2-Acetylpyridine used as flavoring agent. It is also used in application for food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylpyridine used as flavoring agent. It is also used in application for food additive.

Solubility
Soluble in water (18.2 g/100g @ 25C). Soluble in and acetate. Slightly soluble in carbon tetrachloride.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Daniel L. Klayman; Joseph F. Bartosevich; T. Scott Griffin; Carl J. Mason; John P. Scovill. 2-Acetylpyridine thiosemicarbazones. 1. A new class of potential antimalarial agents. J. Med. Chem. 1979, 22(7), 145-150.
  2. John P. Scovill; Daniel L. Klayman; Charles F. Franchino. 2-Acetylpyridine thiosemicarbazones. 4. Complexes with transition metals as antimalarial and antileukemic agents. J. Med. Chem. 1982, 2510), 1261-1264.