4-Chlorophenyl isothiocyanate, 98%
4-Chlorophenyl isothiocyanate, 98%
4-Chlorophenyl isothiocyanate, 98%
Thermo Scientific Chemicals

4-Chlorophenyl isothiocyanate, 98%

CAS: 2131-55-7 | C7H4ClNS | 169.626 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12693.06
also known as A12693-06
Price (USD)
39.65
Special offer
Online exclusive
Ends: 28-Dec-2026
46.50
Save 6.85
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.65
Special offer
Online exclusive
Ends: 28-Dec-2026
46.50
Save 6.85
Each
Chemical Identifiers
CAS2131-55-7
IUPAC Name1-chloro-4-isothiocyanatobenzene
Molecular FormulaC7H4ClNS
InChI KeyMZZVFXMTZTVUFO-UHFFFAOYSA-N
SMILESClC1=CC=C(C=C1)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥97.5%
Melting Point41-47?C
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only