2-Methylbenzyl alcohol, 98%
2-Methylbenzyl alcohol, 98%
2-Methylbenzyl alcohol, 98%
Thermo Scientific Chemicals

2-Methylbenzyl alcohol, 98%

CAS: 89-95-2 | C8H10O | 122.167 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A12696.30
also known as A12696-30
Price (USD)
348.65
Online Exclusive
387.00
Save 38.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
348.65
Online Exclusive
387.00
Save 38.35 (10%)
Each
Chemical Identifiers
CAS89-95-2
IUPAC Name(2-methylphenyl)methanol
Molecular FormulaC8H10O
InChI KeyXPNGNIFUDRPBFJ-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to pale cream
FormCrystals or powder or crystalline powder or fused solid or clear liquid as melt
Assay (GC)≥97.5%
Melting Point (clear melt)31-38°C
2-Methylbenzyl alcohol is widely used strong mobile phase additive in HPLC. it can be used to produce 2-methyl-benzaldehyde at temperature of 20°C. It will need reagent pyridinium chlorochromate with reaction time of 10 min.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbenzyl alcohol is widely used strong mobile phase additive in HPLC. it can be used to produce 2-methyl-benzaldehyde at temperature of 20°C. It will need reagent pyridinium chlorochromate with reaction time of 10 min.

Solubility
Soluble in methanol 0.1 g/mL.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, oxidizing agents.
RUO – Research Use Only

General References:

  1. A Long.; P James.; OP Ward. Aromatic aldehydes as substrates for yeast and yeast alcohol dehydrogenase. Biotechnology and Bioengineering. 1989, 33, (5), 657-660.
  2. PJ Evans.; W Ling.; B Goldschmidt. Metabolites formed during anaerobic transformation of toluene and o-xylene and their proposed relationship to the initial steps of toluene mineralization. Appl. Environ. Microbiol.1992, 58, (2), 496-501.