Acetophenone, 99%
Acetophenone, 99%
Acetophenone, 99%
Thermo Scientific Chemicals

Acetophenone, 99%

CAS: 98-86-2 | C8H8O | 120.151 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 mL
500 mL
2500 mL
Catalog number A12727.0F
also known as A12727-0F
Price (USD)
131.00
Each
Quantity:
2500 mL
Request bulk or custom format
Price (USD)
131.00
Each
Chemical Identifiers
CAS98-86-2
IUPAC Name1-phenylethan-1-one
Molecular FormulaC8H8O
InChI KeyKWOLFJPFCHCOCG-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.5320-1.5350 @20?C
Acetophenone is used as a raw material in the synthesis of active pharmaceutical ingredients such as dextropropoxyphene and phenylpropanolamine. It is employed in organic synthesis and as a polymerization catalyst for olefins. It is involved in the preparation of styrene, 1-phenylethanol. It is also used as a active component of coatings and inks. It reacts with formaldehyde to prepare resins. Further, it is used in textile coatings, in perfumes for orange blossom odor. In addition to this, it is used as a solvent in plastics, adhesives and a flavoring agent in foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetophenone is used as a raw material in the synthesis of active pharmaceutical ingredients such as dextropropoxyphene and phenylpropanolamine. It is employed in organic synthesis and as a polymerization catalyst for olefins. It is involved in the preparation of styrene, 1-phenylethanol. It is also used as a active component of coatings and inks. It reacts with formaldehyde to prepare resins. Further, it is used in textile coatings, in perfumes for orange blossom odor. In addition to this, it is used as a solvent in plastics, adhesives and a flavoring agent in foods.

Solubility
Miscible with water, chloroform, ethanol, ether, benzene and acetone. Slightly miscible with concentrated sulfuric acid.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
RUO – Research Use Only

General References:

  1. Yoshida, H.; Onodera, Y.; Fujita, S.; Kawamori, H.; Arai, M. Solvent effects in heterogeneous selective hydrogenation of acetophenone: differences between Rh/C and Rh/Al2O3 catalysts and the superiority of water as a functional solvent. Green Chem. 2015, 17 (3), 1877-1883.
  2. Villalba, M.; del Pozo, M.; Calvo, E. J. Electrocatalytic hydrogenation of acetophenone and benzophenone using palladium electrodes. Electrochim. Acta 2015, 164, 125-131.