4-Trifluoromethyl-o-phenylenediamine, 98%
4-Trifluoromethyl-o-phenylenediamine, 98%
4-Trifluoromethyl-o-phenylenediamine, 98%
4-Trifluoromethyl-o-phenylenediamine, 98%
Thermo Scientific Chemicals

4-Trifluoromethyl-o-phenylenediamine, 98%

CAS: 368-71-8 | C7H7F3N2 | 176.142 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A12749.03
also known as A12749-03
Price (USD)
41.65
Online Exclusive
45.60
Save 3.95 (9%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
41.65
Online Exclusive
45.60
Save 3.95 (9%)
Each
Add to cart
Chemical Identifiers
CAS368-71-8
IUPAC Name4-(trifluoromethyl)benzene-1,2-diamine
Molecular FormulaC7H7F3N2
InChI KeyRQWJHUJJBYMJMN-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1N)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Pale brown to dark brown or grey
FormCrystals or powder or crystalline powder
Melting Point (clear melt)57.0-63.0°C
3,4-Diaminobenzotrifluoride is commonly used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Diaminobenzotrifluoride is commonly used as pharmaceutical intermediate.

Solubility
Miscible with water.

Notes
Stable at room temperature in closed containers under normal storage and handling conditions. Incompatible to oxidizing agents. Store in a cool, dry place.
RUO – Research Use Only

General References:

  1. R Belcher.;A Sykes.;JC Tatlow. Trifluoromethyl-substituted benziminazoles. J. Chem. Soc. 1954 11, (1) , 4159-4161.
  2. TN Sorrell.; AS Borovik. Absorption, emission, and photophysical properties of copper (I) pyrazole complexes and their carbonyl adducts. Inorganic Chemistry. 1987 26, (12) , 4159-4161.