2-Fluoromesitylene, 98%
2-Fluoromesitylene, 98%
2-Fluoromesitylene, 98%
2-Fluoromesitylene, 98%
Thermo Scientific Chemicals

2-Fluoromesitylene, 98%

CAS: 392-69-8 | C9H11F | 138.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A12779.03
also known as A12779-03
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
58.80
Save 9.15 (16%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
58.80
Save 9.15 (16%)
Each
Add to cart
Chemical Identifiers
CAS392-69-8
IUPAC Name2-fluoro-1,3,5-trimethylbenzene
Molecular FormulaC9H11F
InChI KeyZLGPNBBJPOBSLY-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(F)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4815-1.4855 @ 20?C
Assay (GC)≥97.5%
2-Fluoromesitylene, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoromesitylene, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Incompatible to strong oxidizing agents.
RUO – Research Use Only

General References:

  1. KK Laali.; GI Borodkin. First application of ionic liquids in electrophilic fluorination of arenes; Selectfluor™(F-TEDA-BF 4) for green fluorination. Journal of the Chemical Society. 20022, (1) , 953-957.
  2. GC Finger.; FH Reed.; EW Maynert. Aromatic Fluorine Compounds. III. The Fluoromesitylenes and Derivatives1. Journal of the Chemical Society. 195173, (1) , 149-151.