Pentafluoropropionic acid, 97%
Pentafluoropropionic acid, 97%
Pentafluoropropionic acid, 97%
Thermo Scientific Chemicals

Pentafluoropropionic acid, 97%

CAS: 422-64-0 | C3HF5O2 | 164.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A12791.36
also known as A12791-36
Price (USD)
1,073.65
Online exclusive
1,193.00
Save 119.35
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
1,073.65
Online exclusive
1,193.00
Save 119.35
Each
Chemical Identifiers
CAS422-64-0
IUPAC Namepentafluoropropanoic acid
Molecular FormulaC3HF5O2
InChI KeyLRMSQVBRUNSOJL-UHFFFAOYSA-N
SMILESOC(=O)C(F)(F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index<1.3000 @ 20?C
Appearance (Color)Clear colorless to pink or pale brown
Assay (GC)≥96.0%
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
View more
Pentafluoropropionic acid is used as an efficient catalyst for the preparation of dibenzo[a, j] xanthenes by condensation reaction of beta-naphthol with aryl aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluoropropionic acid is used as an efficient catalyst for the preparation of dibenzo[a, j] xanthenes by condensation reaction of beta-naphthol with aryl aldehydes.

Notes
Hygroscopic. Incompatible withstrong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. The K salt is a convenient alternative to iodopentafluoroethane for the conversion of aryl bromides and iodides to their pentafluoroethyl derivatives, avoiding sealed-tube chemistry: Synth. Commun., 18, 965 (1988):
  2. Yan, H.; Zhang, C.; Zhou, Q.; Yang, S. Occurrence of perfluorinated alkyl substances in sediment from estuarine and coastal areas of the East China Sea. Environ. Sci. Pollut. Res. 2015, 22 (3), 1662-1669.
  3. Zhang, C.; Qu, Y.; Zhao, X.; Zhou, Q. Photoinduced Reductive Decomposition of Perflurooctanoic Acid in Water: Effect of Temperature and Ionic Strength. Clean (Weinh) 2015, 43 (2), 223-228.