2-Methyl-1,3-dioxolane, 98%
2-Methyl-1,3-dioxolane, 98%
2-Methyl-1,3-dioxolane, 98%
Thermo Scientific Chemicals

2-Methyl-1,3-dioxolane, 98%

CAS: 497-26-7 | C4H8O2 | 88.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A12824.09
also known as A12824-09
Price (USD)
56.70
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
56.70
Each
Specifications
Beilstein102520
CAS497-26-7
Chemical Name or Material2-Methyl-1,3-dioxolane
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Density0.982
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only