Chloroacetonitrile, 98+%
Chloroacetonitrile, 98+%
Chloroacetonitrile, 98+%
Chloroacetonitrile, 98+%
Thermo Scientific Chemicals

Chloroacetonitrile, 98+%

CAS: 107-14-2 | C2H2ClN | 75.495 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Catalog number A12825.A1
also known as A12825-A1
Price (USD)
225.65
Online Exclusive
250.00
Save 24.35 (10%)
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)
225.65
Online Exclusive
250.00
Save 24.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS107-14-2
IUPAC Name2-chloroacetonitrile
Molecular FormulaC2H2ClN
InChI KeyRENMDAKOXSCIGH-UHFFFAOYSA-N
SMILESClCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0% (U.K. specification)
Assay from Supplier's CofA≥98.0% (GC) (U.S. specification)
CommentMaterial purchased in the U.S. and in other countries
Identification (FTIR)Conforms (non-U.S. specification)
Refractive Index1.4210-1.4255 @ 20°C (non-U.S. specification)
View more
Chloroacetonitrile is used in the electrochemical synthesis of cyanoacetic acid with carbon dioxide. It is involved in phase-transfer-catalyzed Darzen's condensation reaction with cyclohexanone. It is also used as an eluent additive in thermospray liquid chromatography/mass spectrometry. Further, it is used to prepare polysubstituted pyrido[1,2-a]benzimidazole by reacting with other reactant such as malononitrile, aromatic aldehyde and pyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroacetonitrile is used in the electrochemical synthesis of cyanoacetic acid with carbon dioxide. It is involved in phase-transfer-catalyzed Darzen′s condensation reaction with cyclohexanone. It is also used as an eluent additive in thermospray liquid chromatography/mass spectrometry. Further, it is used to prepare polysubstituted pyrido[1,2-a]benzimidazole by reacting with other reactant such as malononitrile, aromatic aldehyde and pyridine.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and acids.
RUO – Research Use Only

General References:

  1. Forms cyanomethyl esters with carboxylic acids, e.g. in the presence of Et3N, which are a useful protection method; the esters are readily cleaved by Na2S in aqueous acetone: Synth. Commun., 22, 693 (1992).
  2. Good yields of ß-hydroxy nitriles have been obtained by Reformatsky-type reaction with aldehydes and ketones using Zn and TMS chloride: Tetrahedron Lett., 31, 2205 (1990). See Bromoacetonitrile, A13933.
  3. In the presence of base, Darzens condensation with aldehydes and ketones, under conventional or phase-transfer conditions, gives glycidonitriles (cyano epoxides), readily converted to the homologated carboxylic acids, esters or aldehydes with loss of cyanide: J. Chem. Soc., Chem. Commun., 988 (1974); Coll. Czech. Chem. Commun., 53, 822 (1988).
  4. Ritter reaction with tertiary alcohols yields 2-chloroacetamides which, on treatment with thiourea, provides a high-yield conversion of tertiary alcohols to the corresponding amines: Synthesis, 1709 (2000):
  5. For use in vicarious nucleophilic substitution of hydrogen in nitro-substituted hetereocycles, see 2-Nitrothiophene, A17464.
  6. Hoseini, S. J.; Nasrabadi, H.; Fath, R. H.; Moradi, Z.; Rashidi, M. Oxidative Addition of Propargyl Halides, Chloroacetonitrile, and Ethyl Chloroacetate to a Dimethylplatinum(II) Complex: Kinetic and DFT Studies. Organometallics 2014, 33 (7), 1689-1699.
  7. Kimura, S. Y.; Komaki, Y.; Plewa, M. J.; Marinas, B. J. Chloroacetonitrile and N,2-Dichloroacetamide Formation from the Reaction of Chloroacetaldehyde and Monochloramine in Water. Environ. Sci. Technol. 2013, 47 (21), 12382-12390.