4-tert-Pentylcyclohexanone, 97%
4-tert-Pentylcyclohexanone, 97%
4-tert-Pentylcyclohexanone, 97%
Thermo Scientific Chemicals

4-tert-Pentylcyclohexanone, 97%

CAS: 16587-71-6 | C11H20O | 168.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A12832.18
also known as A12832-18
Price (USD)
40.65
Special offer
Online exclusive
Ends: 28-Dec-2026
47.60
Save 6.95
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 28-Dec-2026
47.60
Save 6.95
Each
Chemical Identifiers
CAS16587-71-6
IUPAC Name4-(2-methylbutan-2-yl)cyclohexan-1-one
Molecular FormulaC11H20O
InChI KeyDCSKAMGZSIRJAQ-UHFFFAOYSA-N
SMILESCCC(C)(C)C1CCC(=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
Refractive Index1.4655-1.4705 @ 20?C
FormLiquid
It finds its application as a masking agent and a perfuming agent in cosmetic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a masking agent and a perfuming agent in cosmetic industry.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Shigeaki Okamura, et al. Biological Stereoselective Reduction of 4-Tert-Pentylcyclohexanone By Anthracnose Fungi.Natural Product Letters.,1999,13(4), 313-316.
  2. C.M. Garc, et al. Biotransformations by Colletotrichum species.Tetrahedron: Asymmetry.,2003,14(10), 1229-1239.