Methyl 4,5-dimethoxy-2-nitrobenzoate, 97%
Methyl 4,5-dimethoxy-2-nitrobenzoate, 97%
Methyl 4,5-dimethoxy-2-nitrobenzoate, 97%
Thermo Scientific Chemicals

Methyl 4,5-dimethoxy-2-nitrobenzoate, 97%

CAS: 26791-93-5 | C10H11NO6 | 241.199 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
500 g
Catalog number A12836.36
also known as A12836-36
Price (USD)
238.65
Special offer
Online exclusive
Ends: 30-Jun-2026
280.00
Save 41.35 (15%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
238.65
Special offer
Online exclusive
Ends: 30-Jun-2026
280.00
Save 41.35 (15%)
Each
Chemical Identifiers
CAS26791-93-5
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Melting Point (clear melt)139.0-148.0?C
Appearance (Color)Pale yellow
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only