4-n-Octyloxybenzonitrile, 98%
4-n-Octyloxybenzonitrile, 98%
4-n-Octyloxybenzonitrile, 98%
Thermo Scientific Chemicals

4-n-Octyloxybenzonitrile, 98%

CAS: 88374-55-4 | C15H21NO | 231.339 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12843.22
also known as A12843-22
Price (USD)
347.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
347.00
Each
Chemical Identifiers
CAS88374-55-4
IUPAC Name4-(octyloxy)benzonitrile
Molecular FormulaC15H21NO
InChI KeyGFNSBTARZPEIPN-UHFFFAOYSA-N
SMILESCCCCCCCCOC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Refractive Index1.5080-1.5120 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only