5-Bromovanillin, 98%
5-Bromovanillin, 98%
5-Bromovanillin, 98%
Thermo Scientific Chemicals

5-Bromovanillin, 98%

CAS: 2973-76-4 | C8H7BrO3 | 231.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A12861.14
also known as A12861-14
Price (USD)
26.65
Online exclusive
29.50
Save 2.85
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
26.65
Online exclusive
29.50
Save 2.85
Each
Chemical Identifiers
CAS2973-76-4
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to pale yellow or pale cream to pale brown
FormCrystals or powder or crystalline powder
Melting Point (clear melt)162.0-168.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only