2,4-Dihydroxybenzaldehyde, 98%
2,4-Dihydroxybenzaldehyde, 98%
2,4-Dihydroxybenzaldehyde, 98%
2,4-Dihydroxybenzaldehyde, 98%
Thermo Scientific Chemicals

2,4-Dihydroxybenzaldehyde, 98%

CAS: 95-01-2 | C7H6O3 | 138.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A12868.36
also known as A12868-36
Price (USD)
783.65
Online Exclusive
870.00
Save 86.35 (10%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
783.65
Online Exclusive
870.00
Save 86.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS95-01-2
IUPAC Name2,4-dihydroxybenzaldehyde
Molecular FormulaC7H6O3
InChI KeyIUNJCFABHJZSKB-UHFFFAOYSA-N
SMILESOC1=CC=C(C=O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to pale pink to pale brown
Identification (FTIR)Conforms
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)132.0-139.0?C
FormPowder
2,4-Dihydroxybenzaldehyde is used in two-step synthesis of ethyl 3,5-dibromo-2,4-dihydroxycinnamate. It has a potent antioxidative and antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dihydroxybenzaldehyde is used in two-step synthesis of ethyl 3,5-dibromo-2,4-dihydroxycinnamate. It has a potent antioxidative and antibacterial activity.

Solubility
Soluble in water. Solubility in methanol is almost transparent. Easily soluble in alcohol, ethyl ether, chloroform, glacial acetic acid. Insoluble in benzene and easily decomposes under strong acid strong alkali.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

General References:

  1. Nathalie Gagey, et al. Two-photon uncaging with the efficient 3,5-dibromo-2,4-dihydroxycinnamic caging group.Angew Chem Int Ed Engl.,2007,46(14), 2467-2469.
  2. Mendelson WL, et al. The regioselective 4-benzylation of 2, 4-dihydroxybenzaldehyde.Synth. Commun.,1996,26(3), 593-601.