Water For Injection (WFI) for Cell Culture
Gibco™

Water For Injection (WFI) for Cell Culture

High quality water for use as a solvent in the preparation of cell culture media and laboratory reagents.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1000 mL
10 x 500 mL
6 x 1000 mL
Catalog number A1287301
Price (USD)
27.16
Each
Quantity:
500 mL
Customize this product
Price (USD)
27.16
Each

Gibco™ Water for Injection (WFI) for Cell Culture is high quality water for use as a solvent in the preparation of cell culture media and laboratory reagents. Available in many sizes to meet your needs, Gibco™ Water for Injection (WFI) for Cell Culture features:

  • Water For Injection Quality
  • Gibco™ quality and performance
  • Use in cell culture and molecular biology applications

Water for injection quality

Gibco™ Water for Injection (WFI) for Cell Culture meets the USP monograph for Water for Injection packaged in bulk for commercial use elsewhere. The water is prepared by distillation and filtration (0.1 μm), with no added substances. We monitor the water quality on a routine basis. Conductivity, total organic carbon, endotoxin and bioburden are tested daily; metals are tested monthly.

Gibco™ quality and performance

The performance and consistency expected from Gibco™ liquid media products depends on the high-quality water we use in manufacturing. This water is also bottled as Gibco™ Water for Injection (WFI) for Cell Culture. In addition to meeting USP standards, each lot is quality tested for sterility, endotoxin, conductivity, pH, osmolality, and oxidizable substances.

Use in cell culture and molecular biology applications

Gibco™ Water for Injection (WFI) for Cell Culture can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions and diluting buffers.

Product Use

For Research Use Only or Further Manufacturing. Not for use in diagnostic procedures.

cGMP Manufacturing and Quality System

Gibco™ Water for Injection (WFI) for Cell Culture is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.
Specifications
CAS7732-18-5
Chemical Name or MaterialDihydrogen monoxide
Product LineGibco
Purification MethodDistilled and filtered (0.1 μm)
Recommended StorageStorage conditions: 15 to 30°C, Shipping conditions: Ambient, Shelf life: 24 months from date of manufacture
Shipping ConditionRoom Temperature
SterilitySterile-filtered
Treatment(s)None
GradeCell Culture
Molecular Weight (g/mol)18.02 g/mol
Quantity500 mL
pH4 to 6
Unit SizeEach

Frequently asked questions (FAQs)

What is the difference between Water for Injection (WFI) for Cell Culture, Cat. No. A12873XX and Distilled Water, Cat. No. 15230XXX?

These two products are the same water. However, Water for Injection (WFI) for Cell Culture has undergone more testing, including conductivity measurement as well as testing for nitrate and total organic carbon.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How is Water for Injection (WFI) for Cell Culture prepared?

Water for Injection (WFI) for Cell Culture is prepared by distillation and filtration (0.1 µm), with no added substances. It is labeled “For Research Use Only of Futher Manufacturing. Not for use in diagnostic procedures”.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.