Phenylselenol, 90+%
Phenylselenol, 90+%
Phenylselenol, 90+%
Thermo Scientific Chemicals

Phenylselenol, 90+%

CAS: 645-96-5 | C6H6Se
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A12876.03
also known as A12876-03
Price (USD)
63.65
Online Exclusive
70.50
Save 6.85 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
63.65
Online Exclusive
70.50
Save 6.85 (10%)
Each
Chemical Identifiers
CAS645-96-5
IUPAC Namephenylsulfanyl
Molecular FormulaC6H5Se
InChI KeyYWBHROUQJYHSOR-UHFFFAOYSA-N
SMILES[Se]C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥90.0%
Appearance (Color)Clear colorless to yellow
Refractive Index1.6110-1.6190 @ 20?C
It is applied for reactions of phenylselenol and its anion. Benzeneselenol can used as starting reagent in the synthesis of monoseleno-substituted 1,3-dienes. It may be used in the synthesis of cationic chalcogenolato-bridged diruthenium complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied for reactions of phenylselenol and its anion. Benzeneselenol can used as starting reagent in the synthesis of monoseleno-substituted 1,3-dienes. It may be used in the synthesis of cationic chalcogenolato-bridged diruthenium complexes.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

General References:

  1. Nikolay V Orlov et. al. Exclusive selectivity in the one-pot formation of C-C and C-Se bonds involving Ni-catalyzed alkyne hydroselenation: optimization of the synthetic procedure and a mechanistic study.J Org Chem.,2014,79(24), 12111-12121.
  2. R.J. Puddephatt and P.J. Thompson. Some reactions of methylplatinum and methylgold compounds with phenylselenol, diphenylphosphine, diphenylarsine, N-bromosuccinimide and 2-nitrophenylsulphenyl chloride.J. Organomet. Chem.,1976,117(4), 395-403.
  3. For reactions of phenylselenol and its anion, see Diphenyl diselenide, A13919.