4-Chloroaniline, 98%
4-Chloroaniline, 98%
4-Chloroaniline, 98%
4-Chloroaniline, 98%
Thermo Scientific Chemicals

4-Chloroaniline, 98%

CAS: 106-47-8 | C6H6ClN | 127.571 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A12879.0B
also known as A12879-0B
Price (USD)
119.65
Online Exclusive
133.00
Save 13.35 (10%)
Each
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)
119.65
Online Exclusive
133.00
Save 13.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS106-47-8
IUPAC Name4-chloroaniline
Molecular FormulaC6H6ClN
InChI KeyQSNSCYSYFYORTR-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown or pale yellow
Assay (GC)>97.5%
FormCrystals or crystalline powder or flakes or granules
CommentMay darken on storage
4-Chloroaniline is an important building block used in the chemical industry for the production of drugs and dyestuffs. Some benzodiazepine drugs use 4-chloroaniline in their manufacture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloroaniline is an important building block used in the chemical industry for the production of drugs and dyestuffs. Some benzodiazepine drugs use 4-chloroaniline in their manufacture.

Solubility
Soluble in water (2.6 g/Liter at 20°C ).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides, chloroformates and nitrous acid.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Kathleen E. Simmons.; Robert D. Minard.; Jean Marc Bollag. Oxidative co-oligomerization of guaiacol and 4-chloroaniline. Environ. Sci. Technol. 1989, 23 (1), 115-121.
  2. Donald D. Kaufman.; Jack R. Plimmer.; Ute I. Klingebiel. Microbial oxidation of 4-chloroaniline. J. Agric. Food Chem. 1973, 21 (1), 127-132.