Isopropyltriphenylphosphonium iodide, 98+%
Isopropyltriphenylphosphonium iodide, 98+%
Isopropyltriphenylphosphonium iodide, 98+%
Thermo Scientific Chemicals

Isopropyltriphenylphosphonium iodide, 98+%

CAS: 24470-78-8 | C21H22IP | 432.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
250 g
Catalog number A12881.22
also known as A12881-22
Price (USD)
255.65
Online Exclusive
284.00
Save 28.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
255.65
Online Exclusive
284.00
Save 28.35 (10%)
Each
Chemical Identifiers
CAS24470-78-8
IUPAC Nametriphenyl(propan-2-yl)phosphanium iodide
Molecular FormulaC21H22IP
InChI KeyHHBXWXJLQYJJBW-UHFFFAOYSA-M
SMILES[I-].CC(C)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale yellow
FormCrystalline powder or powder
Elemental AnalysisC: 56.60-60.10% (theory 58.35%)
Water Content (Karl Fischer Titration)≤1.0%
Elemental AnalysisI: 28.48-30.24% (theory 29.36%)
Useful precursor of isopropylidene unit by Wittig reaction with aldehydes. It is also used in tandem cyclopropanation and Wittig olefination in a synthesis of chrysanthemic acid. Also employed in Wittig and cyclopropanation reactions. It is used as a reactant for total synthesis of heliananes, in the preparation of highly substituted benzene derivatives, synthesis of curcuphenol and elvirol analogs via a retro-aldol reaction as fungicidal agents, 4-substituted methoxylbenzoyl-aryl-thiazoles analogues as potent and orally bioavailable anticancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful precursor of isopropylidene unit by Wittig reaction with aldehydes. It is also used in tandem cyclopropanation and Wittig olefination in a synthesis of chrysanthemic acid. Also employed in Wittig and cyclopropanation reactions. It is used as a reactant for total synthesis of heliananes, in the preparation of highly substituted benzene derivatives, synthesis of curcuphenol and elvirol analogs via a retro-aldol reaction as fungicidal agents, 4-substituted methoxylbenzoyl-aryl-thiazoles analogues as potent and orally bioavailable anticancer agents.

Solubility
Solubility in methanol, very faint turbidity.

Notes
Light sensitive and hygroscopic. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Richard A. Bartsch, et. al. Synthesis of highly lipophilic crown ether carboxylic acids.J Org Chem.,1983,48(25), 4864-4869.
  2. Hiroshi Yamataka, et al. Mechanistic study of the Wittig reaction of benzophenone with a nonstabilized ylide.J. Org. Chem.,1988,53(16), 3877-3879.
  3. Useful precursor of isopropylidene unit by Wittig reaction with aldehydes: J. Am. Chem. Soc., 103, 2823 (1981), and ketones: J. Org. Chem., 53, 3877 (1988).
  4. The derived phosphorane can also add to activated double bonds to give gem-dimethyl cyclopropanes: Tetrahedron Lett., 3781 (1972). For use of tandem cyclopropanation and Wittig olefination in a synthesis of chrysanthemic acid, see: Tetrahedron Lett., 3911 (1976):