Benzhydrol, 99%
Benzhydrol, 99%
Benzhydrol, 99%
Thermo Scientific Chemicals

Benzhydrol, 99%

CAS: 91-01-0 | C13H12O | 184.238 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A12884.0B
also known as A12884-0B
Price (USD)
187.65
Online Exclusive
208.00
Save 20.35 (10%)
Each
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)
187.65
Online Exclusive
208.00
Save 20.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS91-01-0
IUPAC Namediphenylmethanol
Molecular FormulaC13H12O
InChI KeyQILSFLSDHQAZET-UHFFFAOYSA-N
SMILESOC(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥98.5%
Melting Point (clear melt)63.0-70.0?C
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Benzhydrol is widely used as intermediates in pharmaceuticals (including antihistamines), agrochemicals, perfumes and other organic compounds. It is used as a fixative in the perfume industry. It is involved in polymerization reaction as a terminating group. It is used as precursor to prepare modafinil, benztropine and diphehydramine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzhydrol is widely used as intermediates in pharmaceuticals (including antihistamines), agrochemicals, perfumes and other organic compounds. It is used as a fixative in the perfume industry. It is involved in polymerization reaction as a terminating group. It is used as precursor to prepare modafinil, benztropine and diphehydramine.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents, acids, acid chlorides and acid anhydrides.
RUO – Research Use Only

General References:

  1. The protection of OH groups as benzhydryl ethers can be catalyzed by sulfuric acid: Org. Synth. Coll., 4, 72 (1963); Angew. Chem. Int. Ed., 15, 281 (1976). See also Benzhydryl bromide, L02211. Thiols, e.g. in cysteine residues in peptide synthesis, can be protected as thioethers in the presence of TFA or HBr/AcOH: J. Chem. Soc. (C), 2683 (1970). The group can be cleaved by TFA in the presence of a cation scavenger, e.g. phenol or anisole (same ref.), or by Na in liquid NH3: J. Am. Chem. Soc., 84, 3887 (1962). See Appendix 6.
  2. Carboxyl groups can be protected by esterification in benzene with tosic acid catalyst. Deprotection can be effected with the same catalyst by refluxing in toluene, which undergoes electrophilic alkylation: Tetrahedron Lett., 37, 1965 (1996).
  3. Misawa, T.; Dodo, K.; Ishikawa, M.; Hashimoto, Y.; Sagawa, M.; Kizaki, M.; Aoyama, H. Structure-activity relationships of benzhydrol derivatives based on 1'-acetoxychavicol acetate (ACA) and their inhibitory activities on multiple myeloma cell growth via inactivation of the NF-κB pathway. Bioorg. Med. Chem. 2015, 23 (9), 2241-2246.
  4. Taskin, O. S.; Temel, B. A.; Tasdelen, M. A.; Yagci, Y. Synthesis of block copolymers by selective H-abstraction and radical coupling reactions using benzophenone/benzhydrol photoinitiating system. Eur. Polym. J. 2015, 62, 304-311.