2-Iodothiophene, 98%, stab. with copper
2-Iodothiophene, 98%, stab. with copper
2-Iodothiophene, 98%, stab. with copper
Thermo Scientific Chemicals

2-Iodothiophene, 98%, stab. with copper

CAS: 3437-95-4 | C4H3IS | 210.032 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A12898.14
also known as A12898-14
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.50
Save 7.85
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.50
Save 7.85
Each
Chemical Identifiers
CAS3437-95-4
IUPAC Name2-iodothiophene
Molecular FormulaC4H3IS
InChI KeyROIMNSWDOJCBFR-UHFFFAOYSA-N
SMILESIC1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow to brown
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.6500-1.6540 @ 20°C
StabilizerCopper powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only