2,5-Dimethoxybenzoic acid, 98+%
2,5-Dimethoxybenzoic acid, 98+%
2,5-Dimethoxybenzoic acid, 98+%
Thermo Scientific Chemicals

2,5-Dimethoxybenzoic acid, 98+%

CAS: 2785-98-0 | C9H10O4 | 182.175 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A12901.22
also known as A12901-22
Price (USD)
477.65
Online Exclusive
531.00
Save 53.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
477.65
Online Exclusive
531.00
Save 53.35 (10%)
Each
Chemical Identifiers
CAS2785-98-0
IUPAC Name2,5-dimethoxybenzoic acid
Molecular FormulaC9H10O4
InChI KeyNYJBTJMNTNCTCP-UHFFFAOYSA-N
SMILESCOC1=CC=C(OC)C(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (Silylated GC)≥98.0%
Melting Point (clear melt)74-80?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only