Barium chloride dihydrate, 99+%
Barium chloride dihydrate, 99+%
Barium chloride dihydrate, 99+%
Barium chloride dihydrate, 99+%
Thermo Scientific Chemicals

Barium chloride dihydrate, 99+%

CAS: 10326-27-9 | BaCl2H4O2 | 244.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A12905.0B
also known as A12905-0B
Price (USD)
108.00
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
108.00
Each
Chemical Identifiers
CAS10326-27-9
IUPAC Namebarium(2+) dihydrate dichloride
Molecular FormulaBaCl2H4O2
InChI KeyPWHCIQQGOQTFAE-UHFFFAOYSA-L
SMILESO.O.[Cl-].[Cl-].[Ba++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale grey
Assay (Titration ex Chloride)≥99.0 to ≤101.0% (non-U.S. specification)
Assay from Supplier's CofA≥99.0% (U.S. specification)
FormCrystalline powder
Solution Test(10% in water) : Clear & colorless (non-U.S. specification)
Barium chloride dihydrate is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Barium chloride dihydrate is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.

Solubility
Soluble in water and metanol. Insoluble in ethanol and ethyl acetate.

Notes
Stable and hygroscopic in nature.
RUO – Research Use Only

General References:

  1. Somroop, K.; Pornprasertsuk, R.; Jinawath, S. Fabrication of Y2O3 -Doped BaZrO3 Thin Films by Electrostatic Spray Deposition. Thin Solid Films 2011, 519 (19), 6408-6412.
  2. Edgar, A.; Zimmermann, J.; von Seggern, H.; Varoy, C. R. Lanthanum-stabilized europium-doped cubic barium chloride: An efficient X-ray phosphor. J. Appl. Phys. 2010, 107 (8), 083516-7.