Reactive Blue 4, dye content min. 35%
Reactive Blue 4, dye content min. 35%
Reactive Blue 4, dye content min. 35%
Thermo Scientific Chemicals

Reactive Blue 4, dye content min. 35%

CAS: 13324-20-4 | C23H14Cl2N6O8S2 | 637.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A12911.22
also known as A12911-22
Price (USD)
122.65
Special offer
Online exclusive
Ends: 30-Jun-2026
144.00
Save 21.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
122.65
Special offer
Online exclusive
Ends: 30-Jun-2026
144.00
Save 21.35 (15%)
Each
Chemical Identifiers
CAS13324-20-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to blue to dark blue
FormPowder
Assay (Dye)Dye content: ≥35%
Solution TestClear blue solution
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only