4-Chloro-1,2-difluorobenzene, 98%
4-Chloro-1,2-difluorobenzene, 98%
4-Chloro-1,2-difluorobenzene, 98%
4-Chloro-1,2-difluorobenzene, 98%
Thermo Scientific Chemicals

4-Chloro-1,2-difluorobenzene, 98%

CAS: 696-02-6 | C6H3ClF2 | 148.537 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12924.14
also known as A12924-14
Price (USD)
124.65
Special offer
Online exclusive
Ends: 30-Jun-2026
146.00
Save 21.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
124.65
Special offer
Online exclusive
Ends: 30-Jun-2026
146.00
Save 21.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS696-02-6
IUPAC Name4-chloro-1,2-difluorobenzene
Molecular FormulaC6H3ClF2
InChI KeyOPQMRQYYRSTBME-UHFFFAOYSA-N
SMILESFC1=CC=C(Cl)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.4720-1.4760 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only