Thioacetamide, 98%
Thioacetamide, 98%
Thioacetamide, 98%
Thermo Scientific Chemicals

Thioacetamide, 98%

CAS: 62-55-5 | C2H5NS | 75.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A12926.22
also known as A12926-22
Price (USD)
65.65
Special offer
Online exclusive
Ends: 30-Jun-2026
77.10
Save 11.45 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
65.65
Special offer
Online exclusive
Ends: 30-Jun-2026
77.10
Save 11.45 (15%)
Each
Chemical Identifiers
CAS62-55-5
IUPAC Nameethanethioamide
Molecular FormulaC2H5NS
InChI KeyYUKQRDCYNOVPGJ-UHFFFAOYSA-N
SMILESCC(N)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)108.0-115.0?C
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Assay (HPLC)≥97.5%
View more
Thioacetamide is used in qualitative inorganic analysis as an in-situ source for sulfide ions. It is used in analytical chemistry as a source of hydrogen sulfide. It is an additive in enantioselective reduction of beta-keto esters with immobilized bakers yeast. It is also used as a vulcanizing agent of polymer and a crosslinking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thioacetamide is used in qualitative inorganic analysis as an in-situ source for sulfide ions. It is used in analytical chemistry as a source of hydrogen sulfide. It is an additive in enantioselective reduction of beta-keto esters with immobilized bakers yeast. It is also used as a vulcanizing agent of polymer and a crosslinking agent.

Solubility
Soluble in ethanol, benzene and petroleum ether. Slightly soluble in water and ether.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. You, M. S.; Lim, C. S.; Kwon, D. H.; Heo, J. H.; Im, S. H.; Chae, K. J. Oxide-free Sb2S3 sensitized solar cells fabricated by spin and heat-treatment of Sb (III)(thioacetamide)2Cl3. Org. Electron. 2015, 21, 155-159.
  2. Wei, D. D.; Wang, J. S.; Li, M. H.; Guo, P. P.; Dong, G.; Yang, M. H.; Kong, L. Y. A pilot study of the onset of hepatic encephalopathy (OHE) in mice induced by thioacetamide and the protective effect of taurine by holistic metabolic characterization. Metabolomics 2015, 11 (3), 559-570.