6-Nitroindole, 97%
6-Nitroindole, 97%
6-Nitroindole, 97%
Thermo Scientific Chemicals

6-Nitroindole, 97%

CAS: 4769-96-4 | C8H6N2O2 | 162.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A12927.06
also known as A12927-06
Price (USD)
338.65
Online Exclusive
376.00
Save 37.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
338.65
Online Exclusive
376.00
Save 37.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS4769-96-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to brown
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)138.0-148.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only