2-Bromopropane, 99%
2-Bromopropane, 99%
2-Bromopropane, 99%
Thermo Scientific Chemicals

2-Bromopropane, 99%

CAS: 75-26-3 | C3H7Br | 122.993 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1000 mL
2500 mL
Catalog number A12944.0F
also known as A12944-0F
Price (USD)
434.65
Online Exclusive
483.00
Save 48.35 (10%)
Each
Quantity:
2500 mL
Request bulk or custom format
Price (USD)
434.65
Online Exclusive
483.00
Save 48.35 (10%)
Each
Chemical Identifiers
CAS75-26-3
IUPAC Name2-bromopropane
Molecular FormulaC3H7Br
InChI KeyNAMYKGVDVNBCFQ-UHFFFAOYSA-N
SMILESCC(C)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.4235-1.4265 @ 20?C
2-Bromopropane serves as an alkylating agent in organic synthesis. It is also used as an intermediate to form alkylated amines and alkylated metallic compounds. Further, it acts as a solvent for industrial cleaning, degreasing, metal processing and finishing, electronics, aerospace and aviation, aerosols, textiles, adhesives and inks. In addition, it is used for introducing the isopropyl functional group in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopropane serves as an alkylating agent in organic synthesis. It is also used as an intermediate to form alkylated amines and alkylated metallic compounds. Further, it acts as a solvent for industrial cleaning, degreasing, metal processing and finishing, electronics, aerospace and aviation, aerosols, textiles, adhesives and inks. In addition, it is used for introducing the isopropyl functional group in organic synthesis.

Solubility
Miscible with alcohol, benzene, chloroform and ether. Slightly miscible with water and acetone.

Notes
Avoid all possible sources of ignition.Incompatible with strong oxidizing agents and strong bases.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Can be used, (K2CO3 in DMF), to protect phenolic OH groups as their isopropyl ethers, more stable than benzyl ethers to Lewis acids: J. Chem. Soc., Perkin 1, 2593 (1979).
  2. Vatsal, A.; Zinjarde, S. S.; Kumar, A. R. Yarrowia lipolytica NCIM 3589, a tropical marine yeast, degrades bromoalkanes by an initial hydrolytic dehalogenation step. Biodegradation 2015, 26 (2), 127-138.
  3. Simonetti, S. O.; Larghi, E. L.; Kaufman, T. S. Alternate and Step-Economic Synthesis of the β-Methylstyrene Chelating Pre-ligand of the Hoveyda-Grubbs’ II Catalyst. Org. Prep. Proced. Int. 2015, 47 (3), 227-231.