2,2-Di-n-propylacetic acid, 98+%
2,2-Di-n-propylacetic acid, 98+%
2,2-Di-n-propylacetic acid, 98+%
Thermo Scientific Chemicals

2,2-Di-n-propylacetic acid, 98+%

CAS: 99-66-1 | C8H16O2 | 144.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A12962.09
also known as A12962-09
Price (USD)
42.00
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
42.00
Each
Chemical Identifiers
CAS99-66-1
IUPAC Name2-propylpentanoic acid
Molecular FormulaC8H16O2
InChI KeyNIJJYAXOARWZEE-UHFFFAOYSA-N
SMILESCCCC(CCC)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4230-1.4270 @ 20?C
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (Silylated GC)≥98.0%
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only