1-Iodohexadecane, 98%, stab. with copper
1-Iodohexadecane, 98%, stab. with copper
1-Iodohexadecane, 98%, stab. with copper
Thermo Scientific Chemicals

1-Iodohexadecane, 98%, stab. with copper

CAS: 544-77-4 | C16H33I | 352.344 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
250 g
Catalog number A12977.30
also known as A12977-30
Price (USD)
218.65
Online Exclusive
243.00
Save 24.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
218.65
Online Exclusive
243.00
Save 24.35 (10%)
Each
Chemical Identifiers
CAS544-77-4
IUPAC Name1-iodohexadecane
Molecular FormulaC16H33I
InChI KeyKMWHQYDMBYABKL-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow or pink
FormLiquid as melt
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4800-1.4840 @ 20?C
View more
1-Iodohexadecane, is used as an ionic liquid for electrochemical applications. It is also used in detection methods as primer and probe design instrumentation applications guide in qPCR Technic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodohexadecane, is used as an ionic liquid for electrochemical applications. It is also used in detection methods as primer and probe design instrumentation applications guide in qPCR Technic.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Keep away from strong oxidizing agents, strong bases. Store it at a temperature of 15 - 30°C. Stable under normal temperatures and pressures.
RUO – Research Use Only

General References:

  1. AK Hajra.; TV Saraswathi.; AK Das. Chemical synthesis of 1-O-alkyl and 1-O-acyl dihydroxyacetone-3-phosphate. Chemistry and Physics of Lipids. 198333, (2), 179-193.
  2. DG Davis.; RJ Kurland. Carbon- 13 NMR of Paramagnetic Iron-Group Cyanides. J. Chem. Phys. 196746, (1) , 388-390.