beta-D-Maltose octaacetate, 98%
beta-D-Maltose octaacetate, 98%
beta-D-Maltose octaacetate, 98%
Thermo Scientific Chemicals

beta-D-Maltose octaacetate, 98%

CAS: 22352-19-8 | C28H38O19 | 678.593 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A12988.06
also known as A12988-06
Price (USD)
99.65
Online Exclusive
111.00
Save 11.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
99.65
Online Exclusive
111.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS22352-19-8
IUPAC Name[4,5,6-tris(acetyloxy)-3-{[3,4,5-tris(acetyloxy)-6-[(acetyloxy)methyl]oxan-2-yl]oxy}oxan-2-yl]methyl acetate
Molecular FormulaC28H38O19
InChI KeyWOTQVEKSRLZRSX-UHFFFAOYNA-N
SMILESCC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC1OC(COC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystalline powder or powder
Assay (unspecified)≥97.5%
Optical Rotation+62.5° ± 2.0° (c=1, chloroform)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only