2',4'-Dimethylacetophenone, 95%
2',4'-Dimethylacetophenone, 95%
2',4'-Dimethylacetophenone, 95%
Thermo Scientific Chemicals

2',4'-Dimethylacetophenone, 95%

CAS: 89-74-7 | C10H12O | 148.205 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A12992.09
also known as A12992-09
Price (USD)
35.65
Online Exclusive
39.70
Save 4.05 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
35.65
Online Exclusive
39.70
Save 4.05 (10%)
Each
Chemical Identifiers
CAS89-74-7
IUPAC Name1-(2,4-dimethylphenyl)ethan-1-one
Molecular FormulaC10H12O
InChI KeyHSDSKVWQTONQBJ-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C)C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5320-1.5370 @ 20°C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only