Sulfanilamide, 98%
Sulfanilamide, 98%
Sulfanilamide, 98%
Thermo Scientific Chemicals

Sulfanilamide, 98%

CAS: 63-74-1 | C6H8N2O2S | 172.202 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A13001.0E
also known as A13001-0E
Price (USD)
324.65
Online exclusive
361.00
Save 36.35
Each
Quantity:
2500 g
Request bulk or custom format
Price (USD)
324.65
Online exclusive
361.00
Save 36.35
Each
Chemical Identifiers
CAS63-74-1
IUPAC Name4-aminobenzene-1-sulfonamide
Molecular FormulaC6H8N2O2S
InChI KeyFDDDEECHVMSUSB-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)S(N)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder or fused solid
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)162.0-168.0°C
Sulfonamide antibacterial Sulfanilamide is an antibacterial and used in the treatment of vaginal yeast infections. It is a competitive inhibitor of dihydropteroate synthestase to block the synthesis of folic acid. It serves as an intermediate for the preparation of 2,6-disubstituted anilines by electrophilic substitution followed by removal of the sulfonamide blocking group by desulfonation with sulfuric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfonamide antibacterial Sulfanilamide is an antibacterial and used in the treatment of vaginal yeast infections. It is a competitive inhibitor of dihydropteroate synthestase to block the synthesis of folic acid. It serves as an intermediate for the preparation of 2,6-disubstituted anilines by electrophilic substitution followed by removal of the sulfonamide blocking group by desulfonation with sulfuric acid.

Solubility
Soluble in water, acetone, ethanol, glycerol, propylene glycol and hydrochloric acid. Insoluble in chloroform, ether, benzene and petroleum ether.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Intermediate for preparation of 2,6-disubstituted anilines, by electrophilic substitution followed by removal of the sulfonamide blocking group by desulfonation with sulfuric acid. See, e.g.: Org. Synth. Coll., 3, 262 (1955).
  2. Yang, L.; Zhou, S.; Xiao, Y.; Tang, Y.; Xie, T. Sensitive simultaneous determination of three sulfanilamide artificial sweeters by capillary electrophoresis with on-line preconcentration and contactless conductivity detection. Food Chem. 2015, 188, 446-451.
  3. Bhattacharya, A.; Sharma, M.; Gulati, A.; Joshi, R., Chanda, S. K.; Ahuja, P. S. Histochemical evaluation of catechins in PEG stressed transgenic tea plants using catechin-specific-diazotized sulfanilamide reagent. Biotech. Histochem. 2015, 90 (1), 45-54.