Tetrabromothiophene, 99%
Tetrabromothiophene, 99%
Tetrabromothiophene, 99%
Thermo Scientific Chemicals

Tetrabromothiophene, 99%

CAS: 3958-03-0 | C4Br4S | 399.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A13009.14
also known as A13009-14
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS3958-03-0
IUPAC Nametetrabromothiophene
Molecular FormulaC4Br4S
InChI KeyAVPWUAFYDNQGNZ-UHFFFAOYSA-N
SMILESBrC1=C(Br)C(Br)=C(Br)S1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)112.0-119.0?C
Appearance (Color)White to pale cream to pale brown
Assay (GC)≥98.5%
FormCrystals or powder or crystalline powder
Tetrabromothiophene, holds wide applications in organic electronics and photonics. It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrabromothiophene, holds wide applications in organic electronics and photonics. It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong bases, strong oxidizing agents. Stable under normal temperatures and pressures.
RUO – Research Use Only

General References:

  1. TT Dang.; N Rasool.; TT Dang.; H Reinke.; P Langer. Synthesis of tetraarylthiophenes by regioselective Suzuki cross-coupling reactions oftetrabromothiophene. Tetrahedron letters. 200748 (5) , 845-847.
  2. A Rahimi.; JC Namyslo.; MHH Drafz. Selective Mono-to Perarylations of Tetrabromothiophene by a Cyclobutene-1, 2-diylbisimidazolium Preligand. J. Org. Chem. 201176 (18) , 7316-7325.