4-(Dimethylamino)pyridine, 99%
4-(Dimethylamino)pyridine, 99%
4-(Dimethylamino)pyridine, 99%
4-(Dimethylamino)pyridine, 99%
Thermo Scientific Chemicals

4-(Dimethylamino)pyridine, 99%

CAS: 1122-58-3 | C7H10N2 | 122.171 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A13016.06
also known as A13016-06
Price (USD)
25.90
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
25.90
Each
Add to cart
Chemical Identifiers
CAS1122-58-3
IUPAC Name4-(dimethylamino)pyridin-1-ium
Molecular FormulaC7H11N2
InChI KeyVHYFNPMBLIVWCW-UHFFFAOYSA-O
SMILESCN(C)C1=CC=[NH+]C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Melting Point (clear melt)109.0-116.0?C
Appearance (Color)White to pale cream
4-(Dimethylamino)pyridine is a useful nucleophilic catalyst for various reactions viz. esterifications with anhydrides, the Baylis-Hillman reaction, hydrosilylations, tritylation and Steglich rearrangement. It is also an effective catalyst for transesterification of beta-keto esters and silylation of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Dimethylamino)pyridine is a useful nucleophilic catalyst for various reactions viz. esterifications with anhydrides, the Baylis-Hillman reaction, hydrosilylations, tritylation and Steglich rearrangement. It is also an effective catalyst for transesterification of beta-keto esters and silylation of alcohols.

Solubility
Soluble in methanol, benzene, ethyl acetate, chloroform, methylene chloride, acetone, and acetic acid. Slightly soluble in ether, diisopropyl ether, cyclohexane, hexane, and water.

Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Hypernucleophilic catalyst. Greatly accelerates the acylation of hindered, including tertiary, alcohols; reviews: Angew. Chem. Int. Ed., 17, 569 (1978). Chem. Soc. Rev., 12, 129 (1983). Catalyst for acylation of alcohols using mixed anhydrides of hindered carboxylic acids with methanesulfonic acid: Synth. Commun., 12, 727 (1982), and with mixed carboxylic anhydrides: J. Org. Chem., 50, 560 (1985). In the Yamaguchi method of macrocyclic lactonization the mixed anhydride formed with 2,4,6-Trichlorobenzoyl chloride, L14159 is cyclized with 2 equiv. of DMAP; high dilution conditions are not required: Bull. Chem. Soc. Jpn., 52, 1989 (1979). For improved method in a high yield synthesis of erythronolide A, see: J. Org. Chem., 55, 7 (1990).
  2. Greatly increases the rate of ester and thio ester formation in the carbodiimide method of coupling (Steglich esterification): Angew. Chem. Int. Ed., 17, 522 (1978). For similar use in macrolactonizations, see: J. Org. Chem., 50, 2394 (1985), and in carbodiimide peptide coupling reactions: Int. J. Pept. Prot. Res., 18, 459 (1981). For peptide reagents, see Appendix 6.
  3. Effective catalyst for transesterification of ß-keto esters in toluene: J. Org. Chem., 50, 3618 (1985), and for smooth decarboalkoxylation of enolizable ß-keto esters in toluene containing a small amount of water: J. Org. Chem., 54, 3474 (1989). Under these conditions, other bases (N,N-dimethylaniline, pyridine, tetra-n-pentylammonium bromide) are ineffective.