3,4-Dimethylphenylhydrazine hydrochloride, 98%
3,4-Dimethylphenylhydrazine hydrochloride, 98%
3,4-Dimethylphenylhydrazine hydrochloride, 98%
3,4-Dimethylphenylhydrazine hydrochloride, 98%
Thermo Scientific Chemicals

3,4-Dimethylphenylhydrazine hydrochloride, 98%

CAS: 60481-51-8 | C8H13ClN2 | 172.656 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A13027.30
also known as A13027-30
Price (USD)
862.65
Online Exclusive
958.00
Save 95.35 (10%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
862.65
Online Exclusive
958.00
Save 95.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS60481-51-8
IUPAC Namehydrogen (3,4-dimethylphenyl)hydrazine chloride
Molecular FormulaC8H13ClN2
InChI KeyYYMIOVAEQIEPET-UHFFFAOYSA-N
SMILES[H+].[Cl-].CC1=CC=C(NN)C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to brown
FormPowder or flakes
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
3,4-Dimethylphenylhydrazine hydrochloride, is used as a reactant with ethyl 3-ethoxy-2-ethoxycarbonylprop-2-enoate to produce 2-(3,4-dimethyl-phenyl)-3-oxo-2,3-dihydro-1H-pyrazole-4-carboxylic acid ethyl ester. This reaction will need reagent K2CO3, and the menstruum ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylphenylhydrazine hydrochloride, is used as a reactant with ethyl 3-ethoxy-2-ethoxycarbonylprop-2-enoate to produce 2-(3,4-dimethyl-phenyl)-3-oxo-2,3-dihydro-1H-pyrazole-4-carboxylic acid ethyl ester. This reaction will need reagent K2CO3, and the menstruum ethanol.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. JA Markwalder.; MR Arnone.; PA Benfield. Synthesis and biological evaluation of 1-aryl-4, 5-dihydro-1 H-pyrazolo [3, 4-d] pyrimidin-4-one inhibitors of cyclin-dependent kinases. J. Med. Chem.200447 (24), 5894-5911.
  2. H Li.; L Cai.; J Li.; Y Hu.; P Zhou.; J Zhang. Novel coumarin fluorescent dyes: Synthesis, structural characterization and recognition behavior towards Cu (II) and Ni (II). Dyes and Pigments. 2011, 91 (3), 309-316.