Phloroglucinol dihydrate, 98%
Phloroglucinol dihydrate, 98%
Phloroglucinol dihydrate, 98%
Phloroglucinol dihydrate, 98%
Thermo Scientific Chemicals

Phloroglucinol dihydrate, 98%

CAS: 6099-90-7 | C6H10O5 | 162.141 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A13034.14
also known as A13034-14
Price (USD)
33.90
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
33.90
Each
Add to cart
Chemical Identifiers
CAS6099-90-7
IUPAC Namebenzene-1,3,5-triol dihydrate
Molecular FormulaC6H10O5
InChI KeyMPYXTIHPALVENR-UHFFFAOYSA-N
SMILESO.O.OC1=CC(O)=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)18-24%
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥97.5%
Appearance (Color)White to cream to brown
Phloroglucinol dihydrate is utilized in the preparation of active pharmaceutical ingredients such as flopropione. It acts as a reagent for the Tollens’ test for pentoses. It is mixed with hydrochloric acid and plays an important role for the detection of lignin (Wiesner test). It is involved in the analysis and qualitative detection of free hydrochloric acid in gastric juice by using Gunzburg reagent. Further, it serves as a coupling agent in printing. It is also employed as a reagent for pentoses, pentosans, aldehydes and as a decalcifier of bone specimens in microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phloroglucinol dihydrate is utilized in the preparation of active pharmaceutical ingredients such as flopropione. It acts as a reagent for the Tollens’ test for pentoses. It is mixed with hydrochloric acid and plays an important role for the detection of lignin (Wiesner test). It is involved in the analysis and qualitative detection of free hydrochloric acid in gastric juice by using Gunzburg reagent. Further, it serves as a coupling agent in printing. It is also employed as a reagent for pentoses, pentosans, aldehydes and as a decalcifier of bone specimens in microscopy.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with acid chlorides, acid anhydrides, bases and oxidizing agents.
RUO – Research Use Only

General References:

  1. Cheng, C.; Liu, C.; Gu, Y. One-pot three-component reactions of methyl ketones, phenols and a nucleophile: an expedient way to synthesize densely substituted benzofurans. Tetrahedron 2015, 71 (42), 8009-8017.
  2. Qin, W.; Lü, H.; Xie, Z. Preparation and evaluation of o-phenanthroline immobilized on a hybrid silica monolith modified with ionic liquids for reversed-phase pressurized capillary electrochromatography. J. Sep. Sci. 2014, 37 (24), 3722-3728.