4-Chlorophenoxyacetic acid, 98+%
4-Chlorophenoxyacetic acid, 98+%
4-Chlorophenoxyacetic acid, 98+%
4-Chlorophenoxyacetic acid, 98+%
Thermo Scientific Chemicals

4-Chlorophenoxyacetic acid, 98+%

CAS: 122-88-3 | C8H7ClO3 | 186.591 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A13065.0E
also known as A13065-0E
Price (USD)
289.65
Special offer
Online exclusive
Ends: 30-Jun-2026
340.00
Save 50.35 (15%)
Each
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)
289.65
Special offer
Online exclusive
Ends: 30-Jun-2026
340.00
Save 50.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS122-88-3
IUPAC Name2-(4-chlorophenoxy)acetic acid
Molecular FormulaC8H7ClO3
InChI KeySODPIMGUZLOIPE-UHFFFAOYSA-N
SMILESOC(=O)COC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Melting Point (clear melt)155.0-161.0?C
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only