2-(2-Chloroethoxy)ethanol, 99%
2-(2-Chloroethoxy)ethanol, 99%
2-(2-Chloroethoxy)ethanol, 99%
Thermo Scientific Chemicals

2-(2-Chloroethoxy)ethanol, 99%

CAS: 628-89-7 | C4H9ClO2 | 124.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A13074.0B
also known as A13074-0B
Price (USD)
510.65
Online Exclusive
567.00
Save 56.35 (10%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
510.65
Online Exclusive
567.00
Save 56.35 (10%)
Each
Chemical Identifiers
CAS628-89-7
IUPAC Name2-(2-chloroethoxy)ethan-1-ol
Molecular FormulaC4H9ClO2
InChI KeyLECMBPWEOVZHKN-UHFFFAOYSA-N
SMILESOCCOCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Clear colorless
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.4520-1.4550 @ 20?C
2-(2-Chloroethoxy)ethanol is employed in the preparation of o-nitrophenylbromoacetaldehyde bis-2-(2-chloroethoxy)-ethyl acetal, quetiapine and 2-(2-azidoethoxy)ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Chloroethoxy)ethanol is employed in the preparation of o-nitrophenylbromoacetaldehyde bis-2-(2-chloroethoxy)-ethyl acetal, quetiapine and 2-(2-azidoethoxy)ethanol.

Solubility
Miscible with water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Kotb, E. R.; Abbas, H. A. S.; Flefel, E. M.; Sayed, H. H.; Abdelwahed, N. A. M. Month 2015 Utility of Hantzsch Ester in Synthesis of Some 3,5-Bis-dihydropyridine Derivatives and Studying Their Biological Evaluation. J. Heterocycl. Chem. 2015, 52 (5), 1531-1539.
  2. Yang, Y.; Fu, H.; Cui, M.; Peng, C.; Liang, Z.; Dai, J.; Zhang, Z.; Lin, C.; Liu, B. Preliminary evaluation of fluoro-pegylated benzyloxybenzenes for quantification of beta-amyloid plaques by positron emission tomography. Eur. J. Med. Chem. 2015, 104, 86-96.