2-Methylphenoxyacetic acid, 98%
2-Methylphenoxyacetic acid, 98%
2-Methylphenoxyacetic acid, 98%
Thermo Scientific Chemicals

2-Methylphenoxyacetic acid, 98%

CAS: 1878-49-5 | C9H10O3 | 166.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A13076.30
also known as A13076-30
Price (USD)
810.65
Online Exclusive
901.00
Save 90.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
810.65
Online Exclusive
901.00
Save 90.35 (10%)
Each
Chemical Identifiers
CAS1878-49-5
IUPAC Name2-(2-methylphenoxy)acetic acid
Molecular FormulaC9H10O3
InChI KeyQJVXBRUGKLCUMY-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1OCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (GC)≥97.5%
Melting Point (clear melt)152.0-158.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only