2-Mercapto-1-methylimidazole, 98%
2-Mercapto-1-methylimidazole, 98%
2-Mercapto-1-methylimidazole, 98%
2-Mercapto-1-methylimidazole, 98%
Thermo Scientific Chemicals

2-Mercapto-1-methylimidazole, 98%

CAS: 60-56-0 | C4H6N2S | 114.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A13094.14
also known as A13094-14
Price (USD)
53.65
Online Exclusive
59.50
Save 5.85 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
59.50
Save 5.85 (10%)
Each
Add to cart
Chemical Identifiers
CAS60-56-0
IUPAC Name1-methyl-2,3-dihydro-1H-imidazole-2-thione
Molecular FormulaC4H6N2S
InChI KeyPMRYVIKBURPHAH-UHFFFAOYSA-N
SMILESCN1C=CNC1=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Melting Point (clear melt)141.0-147.0?C
Assay (GC)≥97.5%
Possesses antioxidant properties

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only