Adipoyl chloride, 98%
Adipoyl chloride, 98%
Adipoyl chloride, 98%
Adipoyl chloride, 98%
Thermo Scientific Chemicals

Adipoyl chloride, 98%

CAS: 111-50-2 | C6H8Cl2O2 | 183.028 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A13168.22
also known as A13168-22
Price (USD)
154.65
Special offer
Online exclusive
Ends: 30-Jun-2026
182.00
Save 27.35 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
154.65
Special offer
Online exclusive
Ends: 30-Jun-2026
182.00
Save 27.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS111-50-2
IUPAC Namehexanedioyl dichloride
Molecular FormulaC6H8Cl2O2
InChI KeyPWAXUOGZOSVGBO-UHFFFAOYSA-N
SMILESClC(=O)CCCCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown to dark brown
Assay (GC)≥97.5%
Refractive Index1.4670-1.4730 @ 20?C
FormLiquid
CommentMay darken on storage
Adipoyl chloride is used in the production of nylon by reacting with hexamethylene diamine. It is also used in the preparation of biphenyl end-capped liquid crystals. Further, it is employed in the synthesis of chiral polymer for membrane applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adipoyl chloride is used in the production of nylon by reacting with hexamethylene diamine. It is also used in the preparation of biphenyl end-capped liquid crystals. Further, it is employed in the synthesis of chiral polymer for membrane applications.

Solubility
Immiscible with water.

Notes
Incompatible strong oxidizing agents, bases and acids.
RUO – Research Use Only

General References:

  1. Al Tamimi, I. O.; Al Heeti, M. I.; Aziz, J. Z. Preparation of Dioxadiazole from Aryl Acid Hydrazide with Adipoyl Chloride. Int. J. Curr. Microbiol. App. Sci. 2015, 4 (12), 630-639.
  2. Dasgupta, Q.; Chatterjee, K.; Madras, G. Controlled Release of Salicylic Acid from Biodegradable Cross-Linked Polyesters. Mol. Pharmaceutics 2015, 12 (9), 3479-3489.