Naphthalene, 99+%
Naphthalene, 99+%
Naphthalene, 99+%
Thermo Scientific Chemicals

Naphthalene, 99+%

CAS: 91-20-3 | C10H8 | 128.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A13188.0B
also known as A13188-0B
Price (USD)
58.70
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
58.70
Each
Chemical Identifiers
CAS91-20-3
IUPAC Namenaphthalene
Molecular FormulaC10H8
InChI KeyUFWIBTONFRDIAS-UHFFFAOYSA-N
SMILESC1=CC2=CC=CC=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder or fused solid or flakes
Appearance (Color)White
Assay (GC)≥99.0%
Melting Point (clear melt)78.0-84.0?C
Naphthalene is formerly important in dyestuff manufacture. It acts as a precursor to phthalic anhydride. Naphthalene is used in the manufacturing of phthalic anhydride, naphthalene sulfonic acids and hydrogenated naphthalenes such as 1,2,3,4-tetrahydronaphthalene, which is used as a precursor for various dyestuffs, pigments, rubber processing chemicals and in pharmaceuticals. Its molten form is used as solvent replacing the high- boiling solvents like dichlorobenzene, nitrobenzene and durene. It is involved as reaction medium for in the Diels-Alder reaction between C60 with anthracene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Naphthalene is formerly important in dyestuff manufacture. It acts as a precursor to phthalic anhydride. Naphthalene is used in the manufacturing of phthalic anhydride, naphthalene sulfonic acids and hydrogenated naphthalenes such as 1,2,3,4-tetrahydronaphthalene, which is used as a precursor for various dyestuffs, pigments, rubber processing chemicals and in pharmaceuticals. Its molten form is used as solvent replacing the high- boiling solvents like dichlorobenzene, nitrobenzene and durene. It is involved as reaction medium for in the Diels-Alder reaction between C60 with anthracene.

Solubility
Soluble in benzene, toluene, chloroform, carbon disulfide, ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Reacts with Na metal in 1,2-dimethoxyethane to give Na naphthalenide. This radical anion is a powerful-electron donor, e.g. in reductive dehalogenation of vic-dihalides: J. Chem. Soc., Chem. Commun., 78 (1969), and in reductive cleavage of tosylates to alcohols: J. Am. Chem. Soc., 88, 1581 (1966).
  2. Li naphthalenide is a useful, non-nucleophilic reagent for the conversion of carboxylic acids to their dianions in cases where LDA could interfere: J. Org. Chem., 45, 1106 (1980). Similarly, has been found preferable to LDA in the lithiation of allylic nitriles: Synthesis, 358 (1981).
  3. Carboxylic acids react with alkyl, alkenyl or aryl chlorides in the presence of Li metal and a catalytic amount of naphthalene in THF to give ketones in fair to good yields: J. Org. Chem., 61, 6058 (1996).
  4. Hwang, Y. J.; Earmme, T.; Courtright, B. A.; Eberle, F. N.; Jenekhe, S. A. n-Type Semiconducting Naphthalene Diimide-Perylene Diimide Copolymers: Controlling Crystallinity, Blend Morphology, and Compatibility Toward High-Performance All-Polymer Solar Cells. J. Am. Chem. Soc. 2015, 137 (13), 4424-4434.
  5. Attah, I. K.; Platt, S. P.; Meot-Ner, M.; El-Shall, M. S.; Peverati, R.; Head-Gordon, M. What Is the Structure of the Naphthalene-Benzene Heterodimer Radical Cation? Binding Energy, Charge Delocalization, and Unexpected Charge-Transfer Interaction in Stacked Dimer and Trimer Radical Cations. J. Phys. Chem. Lett. 2015, 6 (7), 1111-1118.