1,3-Dichloro-5-iodobenzene, 99%
1,3-Dichloro-5-iodobenzene, 99%
1,3-Dichloro-5-iodobenzene, 99%
Thermo Scientific Chemicals

1,3-Dichloro-5-iodobenzene, 99%

CAS: 3032-81-3 | C6H3Cl2I | 272.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A13192.06
also known as A13192-06
Price (USD)
37.65
Special offer
Online exclusive
Ends: 30-Jun-2026
44.30
Save 6.65 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
37.65
Special offer
Online exclusive
Ends: 30-Jun-2026
44.30
Save 6.65 (15%)
Each
Chemical Identifiers
CAS3032-81-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to pale brown or green
FormCrystalline powder or powder or lumpy powder
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)53-61°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only