2-Methoxynicotinic acid, 98%
2-Methoxynicotinic acid, 98%
2-Methoxynicotinic acid, 98%
2-Methoxynicotinic acid, 98%
Thermo Scientific Chemicals

2-Methoxynicotinic acid, 98%

CAS: 16498-81-0 | C7H6NO3 | 152.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A13193.03
also known as A13193-03
Price (USD)
19.30
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
19.30
Each
Add to cart
Chemical Identifiers
CAS16498-81-0
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (GC)≥96.0%
Melting Point147-153°C
Appearance (Color)White to cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only