2-Methylbenzoxazole, 97%
2-Methylbenzoxazole, 97%
2-Methylbenzoxazole, 97%
2-Methylbenzoxazole, 97%
Thermo Scientific Chemicals

2-Methylbenzoxazole, 97%

CAS: 95-21-6 | C8H7NO | 133.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A13198.14
also known as A13198-14
Price (USD)
50.65
Special offer
Online exclusive
Ends: 30-Jun-2026
59.70
Save 9.05 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
50.65
Special offer
Online exclusive
Ends: 30-Jun-2026
59.70
Save 9.05 (15%)
Each
Add to cart
Chemical Identifiers
CAS95-21-6
IUPAC Name2-methyl-1,3-benzoxazole
Molecular FormulaC8H7NO
InChI KeyDQSHFKPKFISSNM-UHFFFAOYSA-N
SMILESCC1=NC2=CC=CC=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5475-1.5525 @ 20°C [non-U.S. sourced material]
Assay from Supplier's CofA≥96.0% [U.S sourced material]
CommentMaterial sourced in the U.S. and in other countries
Identification (FTIR)Conforms [non-U.S. sourced material]
Proton NMRConforms to structure [U.S. sourced material]
View more
2-Methylbenzoxazole has been used in synthesis of bis-styryl dyes. It is also used in medicine and for the synthesis of other organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbenzoxazole has been used in synthesis of bis-styryl dyes. It is also used in medicine and for the synthesis of other organic compounds.

Solubility
Not miscible in water.

Notes
It is sensitive to light. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Ioannis N.Houpis; Audrey Molina; Joseph Lynch; Robert A.Reamer; R.P.Volante, Paul J.Reider. Condensation of 2-methylbenzoxazole with aromatic aldehydes bearing acidic protons. A convenient coupling in the synthesis of the HIV-reverse transcriptase inhibitor L-696,229. J. Org. Chem. 1993, 58, (11),3176-3178
  2. M.Massacesi; R.Pinna; M.Biddau; G.Ponticelli; I.A.Zakharova. Spectroscopic and biological characterization of the palladium(II) and platinum(II) complexes with benzoxazole and 2-methylbenzoxazole. Inorganica Chimica Acta. 1983, 80, 151-155