3-Chlorobenzonitrile, 99%
3-Chlorobenzonitrile, 99%
3-Chlorobenzonitrile, 99%
Thermo Scientific Chemicals

3-Chlorobenzonitrile, 99%

CAS: 766-84-7 | C7H4ClN | 137.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A13237.09
also known as A13237-09
Price (USD)
55.65
Online Exclusive
61.80
Save 6.15 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
55.65
Online Exclusive
61.80
Save 6.15 (10%)
Each
Chemical Identifiers
CAS766-84-7
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps or fused solid
Water Content (Karl Fischer Titration)≤0.75%
Appearance (Color)White to pale cream to pale yellow to pale brown
Assay (GC)≥98.5%
Melting Point (clear melt)36.0-43.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only